Would you like to log in again?
MPure Cultured Cell DNA Extraction Kit, MP Biomedicals
Supplier: MP Biomedicals
MPure™
117022500 117022500
10204-732EA 605.84 USD InStock
10204-732 CA76046-200
MPure Cultured Cell DNA Extraction Kit, MP Biomedicals
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MPure Cultured Cell DNA Extraction Kit is used with the MPure-12 Nucleic Acid Purification System for the extraction and purification of genomic DNA from cultured cells and buffy coats. Nucleic acids extracted and purified from MPure Cultured Cell DNA Extraction Kit can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis.
  • Extraction of genomic DNA from cultured cells in monolayer or suspension and from buffy coat
  • Completely automated purification protocol
  • Use with MPure-12 System
  • Run in as little as 40 minutes
  • Nucleic Acids can be used immediately in downstream applications
Nucleic acids extracted and purified from MPure Cultured Cell DNA Extraction Kit can be used in a number of downstream applications including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP and Southern Blot Analysis.
Capacity: Do not use more than 5 x 106 cells with a normal set of chromosomes
Packaging: Components Proteinase K solution = 40 µL, Lysis Buffer 2 = 720 µL, Binding Buffer 1 = 720 µL, Magnetic Bead Solution = 800 µL, Washing Buffer 1 = 1000 µL, Washing Buffer 2 = 1000 µL, Washing Buffer 3 = 1000 µL, Elution Buffer 1 = 1000 µL, Elution Buffer 2 = 1000 µL
Caution: Store at room temperature (15-25°C). Do not freeze the reagent cartridges. The Kits are stable for 12 months under these conditions. Store the purified DNA at 4°C (short-term) or aliquot and store at –70°C (long-term) before performing the downstream analysis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty