Would you like to log in again?
MPure FFPE DNA Extraction Kit, MP Biomedicals, LLC
Supplier: MP Biomedicals
MPure™
117022900 117022900
10204-736EA 605.84 USD InStock
CA76046-204 10204-736
MPure FFPE DNA Extraction Kit, MP Biomedicals, LLC
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
MPure FFPE DNA Extraction Kit is used with the MPure-12 Nucleic Acid Purification aNAP System for the extraction and purification of genomic DNA from FFPE (Formalin-Fixed, Paraffin-Embedded) tissue samples
  • Extraction of genomic DNA from Formalin-Fixed, Paraffin-Embedded (FFPE) tissue samples
  • Completely automated purification protocol
  • Use with MPure-12 System
  • Run in as little as 35 minutes
  • Nucleic Acids can be used immediately in downstream applications
Nucleic acids extracted and purified from MPure FFPE DNA Extraction Kit can be used in a number of downstream application including: PCR; qPCR; Sequencing (NGS); Microarray; RFLP and; Southern Blot Analysis.
Capacity: 100-400 µL/ One to eight 10 µm- thick sections of tissue. 100-400 µL/ three to ten biopsies.
Packaging: Components Lysis Buffer 2 = 720 µL, Binding Buffer 1 = 720 µL, Magnetic Bead Solution = 800 µL, Washing Buffer 1 = 1000 µL, Washing Buffer 2 = 1000 µL, Washing Buffer 3 = 1000 µL, Elution Buffer 1 = 1000 µL, Elution Buffer 2 = 1000 µL
Caution: Store at room temperature (15-25°C). Do not freeze the reagent cartridges. The Kits are stable for 12 months under these conditions. Store the purified DNA at 4°C (short-term) or aliquot and store at -70°C (long-term) before performing the downstream analysis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty