Would you like to log in again?
Ward's® Blood Comparison Set
Supplier: Avantor
WARD470178-346
470178-346KT 52.25 USD InStock
470178-346
Ward's® Blood Comparison Set
Slides Prepared Slides
Introduce Your Students to the Surprising Diversity of Animal Blood Morphology.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Different species have different components and even different colors than what students may expect. With this slide set, students will be able to observe these many differences in the blood slides included in this kit. They will then be able to identify characteristics particular to each animal and observe the adaptations that allow each animal to survive in its respective environment.
Ordering information: Set includes human, cat, camel, bird, fish, frog, and horseshoe crab blood smears. An instructional booklet is also included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty