Would you like to log in again?
Specialized Clamps and Accessories, Thermo Scientific
Supplier: Thermo Scientific
176010 715030 176010 715030
16002-302EA 236.86 USD InStock
CA84000-482 CA16002-302 16002-302 84000-482
Specialized Clamps and Accessories, Thermo Scientific
Clamps General Purpose Clamps
Ensure accuracy and efficiency with Thermo Scientific™ Specialized Clamps and Accessories.
  • For clamping thin materials
  • Spring-loaded jaws secure specimen
  • Compensation slot ensures correct specimen clamping
  • Orienting on the Z-axis
  • Foil clamp is fitted directly on the specimen cylinder
  • Sandwich supporting material is for foil clamp to section thin foil specimens
  • 50 squares of hard material and two strips
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty