Would you like to log in again?
ExoQuant™ Overall Exosome Capture and Quantification Assay Kit (Human Serum, Colorimetric), BioVision
Supplier: BioVision
AB287861-96TEST
75799-372EA 1581.35 USD InStock
75799-372
ExoQuant™ Overall Exosome Capture and Quantification Assay Kit (Human Serum, Colorimetric), BioVision
Assays Cellular Assays
Ensure dose-dependent exosome detection in a variety of samples.
  • Enables the calculation of the approximate exosome quantity
  • Load up to 100 µl of biological sample
  • Ensure dose-dependent exosome detection
ExoQuant™ Kits ensure dose-dependent exosome detection in a variety of samples. The quantification can be both relative and absolute as the assay calibration enables the calculation of the approximate exosome quantity for each sample. Exosome comprehensive profiling. Up to 100 µl of biological sample can be loaded per well. Unfractionated serum sample can be directly used for capture.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty