Would you like to log in again?
TotalExoRNA™Overall Exosome Immunocapture and RNA Extraction Kit (Biological Fluids/Cell Media), BioVision Inc
Supplier: BioVision
AB287871-20TEST
75799-390EA 1817.33 USD InStock
75799-390
TotalExoRNA™Overall Exosome Immunocapture and RNA Extraction Kit (Biological Fluids/Cell Media), BioVision Inc
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The only kit on the market providing exosome standards as control.
  • Direct capture and exosome RNA extraction from human biofluids
  • Whole plasma and serum can be directly used for capture
  • Better RNA yield than similar products
Direct capture and exosome RNA extraction from human biofluids and cell culture media without initial exosome purification step. Starting from 0.1 ml of biological sample (plasma) per reaction. Whole plasma and serum can be directly used for capture. Concentrated urine and cell culture supernatant samples are recommended prior capture according to our suggested protocol, Simultaneous miRNA and mRNA profiling (qRT-PCR, RT-PCR, microarray). Fast and user friendly protocol. Small starting amount of sample (less than 1 ml). Better RNA yield than similar products. The only kit on the market providing exosome standards as control. RNA extraction from overall exosome population captured with immunobeads (from human biofluids and cell media).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty