Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BasicExoRNA™Basic RNA Extraction Kit (Pre-isolated Exosomes),BioVision Inc.
Supplier: BioVision
BasicExoRNA™
K1222-25 AB287869-50TEST
75799-396EA 675.94 USD InStock
75799-396 75799-398
BasicExoRNA™Basic RNA Extraction Kit (Pre-isolated Exosomes),BioVision Inc.
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The only kit on the market providing exosome standards as control.
  • RNA isolation from pre-isolated exosome
  • Better RNA yield than similar products
  • Fast and user friendly protocol
Fast and user friendly protocol. Small starting amount of sample (less than 1 ml). Better RNA yield than similar products. The only kit on the market providing exosome standards as control. RNA isolation from pre-isolated exosomes isolated via ultracentrifuge, chemical precipitation, size chromatography, immunocapture, etc.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty