Would you like to log in again?
ExoFACS™ Kit for Cell Culture Media Exosomes, BioVision
Supplier: BioVision
AB287876-20TEST
75799-464EA 1274.45 USD InStock
75799-464
ExoFACS™ Kit for Cell Culture Media Exosomes, BioVision
Protein Extraction Protein Extraction Kits
Exosome isolation from biofluids or cell culture media and FACS analysis of exosome markers.
  • Exosome comprehensive profiling
  • User-friendly
  • No initial exosome purification required
Exosome comprehensive profiling. User friendly and suitable for multiple marker analyses. No initial exosome purification required. Lyophilized Exosome Standards for positive control included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty