Would you like to log in again?
Adson-Brown Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
47-2447
89024-980EA 170.48 USD InStock
89024-980
Adson-Brown Tissue Forceps, OR Grade, Sklar
Forceps
Sklar® Adson-Brown Tissue Forceps are designed for holding and manipulating delicate tissues
Censitrac Ready products are industry standard and constructed of stainless steel.
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty