Would you like to log in again?
Arizant Bair Hugger™ Dual Torso Warming Blanket, 3M™
Supplier: 3M
Arizant Bair Hugger™
MMM54200
75787-596CS 542.73 USD InStock
75787-596
Arizant Bair Hugger™ Dual Torso Warming Blanket, 3M™
First Aid Supplies Surgical Drapes
This forced-air warming blanket is specifically designed for use during lower body surgical procedures.
  • Effectively used for patients in the supine, lateral, and other positions
  • Two convenient, resealable hose ports
  • Pre-attached clear head drape retains warm air around patient's head and allows observation by the clinician
  • Integrated tuck flaps at shoulders help maintain blanket position
  • Soft, comfortable, lightweight, latex-free material
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty