Would you like to log in again?
Arizant Bair Paws™ Pediatric Warming Gowns, 3M™
Supplier: 3M
MMM83501 MMM84501 MMM81501 MMM83101
75786-958CS 1070.18 USD InStock
75786-958 75786-956 75786-966 75786-950
Arizant Bair Paws™ Pediatric Warming Gowns, 3M™
Disposable Garments
Arizant Bair Paws™ system has revolutionized forced-air warming by providing clinicians options for comfort and clinical warming throughout the perioperative process in one convenient gown.
  • Booties, in conjunction with the gown, help keep the patient comfortable
  • Refastenable strips provide quick access to the arms and chest
  • Side ties ensure full, secure coverage, even in the back
  • Dual air-channel inserts and hose ports enable prewarming and comfort warming
  • Soft material ensures greater personal comfort
  • Convenient and easy to use
Dual air-channel inserts and hose ports enable prewarming and comfort warming with a Bair Paws™ 800 series warming unit, or clinical warming with a Bair Hugger™ 500 or 700 series temperature management unit (the Bair Paws™ hose port is on the lower right side; the Bair Hugger™ hose port is on the upper left shoulder.)
The Bair Paws™ system offers a pediatric size, providing a way to warm and comfort some of the smallest surgical patients.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty