Would you like to log in again?
Introductory Molecular Model Set
Supplier: CHRISTENSEN EDUC MATERIALS SE
PE7078
470200-164EA 39.3 USD InStock
470200-164
Introductory Molecular Model Set
Models Molecular Biology Models
Economical Kit Ideal for Individual Student Use.
  • Wooden Parts
  • Springs represent bonds
Start students off simply, with the basic components of organic molecular structure. Pre-drilled wooden balls represent atoms, while three different lengths of springs represent different types of molecular bonds. The set comes with instructions; 30 springs 1", 1 1/8", or 1 1/2" long; and 35 pre-drilled, colored wooden balls 3/4" in diameter: six carbon (black), two nitrogen (blue), 15 hydrogen (yellow), four oxygen (red), four chlorine (green), two bromine (orange), and two iodine/fluorine (purple).
Caution: WARNING: CHOKING HAZARD–Small Parts. Not for children under 3 yrs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty