Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Feaster Wire Speculum, OR Grade, Sklar
Supplier: Sklar
65-1760
10841-340EA 103.59 USD InStock
10841-340
Feaster Wire Speculum, OR Grade, Sklar
Specula
This is an ophthalmic instrument used to retain the eyelids open, often during cataract or phacoemulsification procedures.
  • Made of stainless steel
It is utilized to provide the surgeon with increased visualization and access to the surgical site. This speculum is lightweight and does not put a great deal of pressure on the globe. To a degree, the surgeon can increase the tension of the instrument by manipulating the wire where the speculum is held, as it is slightly maleable. In situations where a wire speculum will not provide enough force to open the lids sufficiently, a speculum with a screw mechanism may be used.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty