Would you like to log in again?
Crile-Wood Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-1280 20-1295 20-1270 20-1265
82030-092EA 186.63 USD InStock
82030-092 82030-090 82030-098 82030-094
Crile-Wood Needle Holder, OR Grade, Sklar
Forceps
Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials.
The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders.The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty