Would you like to log in again?
Arizant Bair Hugger™ Full Body Warming Blanket, 3M™
Supplier: 3M
Arizant Bair Hugger™
MMM30000
75787-568CS 258.05 USD InStock
75787-568
Arizant Bair Hugger™ Full Body Warming Blanket, 3M™
First Aid Supplies Surgical Drapes
This forced-air warming blanket provides coverage to the entire patient.
  • Facilitates maximum thermal transfer with a blanket that hugs the patient
  • Foot drape minimizes risk of thermal injury to the feet and lower leg areas
  • Integrated tuck flaps at shoulders help maintain blanket position
  • Soft, comfortable, lightweight, latex-free material
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty