Would you like to log in again?
Surgi-OR™ TC Iris Scissors, Physician Grade, Sklar
Supplier: Sklar
95-124 95-125
89023-478EA 205.16 USD InStock
89023-478 89023-476
Surgi-OR™ TC Iris Scissors, Physician Grade, Sklar
Scissors
Originally designed for ophthalmic procedures, Iris Scissors have become a multi-purpose instrument used in various surgical environments. They provide reliable instrumentation for the operating room and other surgical centers.
These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Hand-crafted in Pakistan, Surgi-OR™ TC Iris Scissors are a mid-grade instrument forged from the highest quality Japanese steel. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards. Tungsten Carbide instruments are designed to stay sharp for a significantly longer period of time than other standard instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty