Would you like to log in again?
Surgi-OR™ TC Strabismus Scissors, Physician Grade, Sklar
Supplier: Sklar
95-127 95-126
89023-480EA 196.13 USD InStock
89023-480 89023-482
Surgi-OR™ TC Strabismus Scissors, Physician Grade, Sklar
Scissors
Surgi-OR™ TC Strabismus Scissors are an ophthalmic dissecting instrument primarily used during procedures for the treatment of Strabismus, otherwise known as "cross-eyes".
Hand-crafted in Pakistan, these mid-grade instruments are forged from the highest quality Japanese steel. They provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. Surgi-OR™ instruments are a cost-effective line of instruments that must meet Sklar's quality standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty