Would you like to log in again?
Surgi-OR™ Lister Bandage Scissors, Physician Grade, Sklar
Supplier: Sklar
95-222 95-231 95-241
89023-528EA 45.59 USD InStock
89023-528 89023-524 89023-532
Surgi-OR™ Lister Bandage Scissors, Physician Grade, Sklar
Scissors
Surgi-OR™ Lister Bandage Scissors are a versatile instrument most commonly used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric, and occasionally umbilical cord.
The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty