Would you like to log in again?
Surgi-OR™ Nurse's Bandage Scissors, Physician Grade, Sklar
Supplier: Sklar
95-232
89023-530EA 17.89 USD InStock
89023-530
Surgi-OR™ Nurse's Bandage Scissors, Physician Grade, Sklar
Scissors
Surgi-OR™ Nurse's Bandage Scissors are non-ratcheted, finger ring scissors primarily used for removing tight bandages.
The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. The shanks are straight and feature a pocket clip. After the screw lock the blades angle slightly.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty