Would you like to log in again?
Curos Jet™ Disinfecting Caps, 3M™
Supplier: 3M
MMMCFJ5-250
75787-628CS 4653.28 USD InStock
75787-628
Curos Jet™ Disinfecting Caps, 3M™
Closures Safety Caps and Safety Waste Caps
Curos Jet™ disinfecting caps are the next-generation solution for I.V.
  • Caps twist on easily and stay on commonly-used needleless connectors
  • Caps are packaged to hang from an I.V. pole for easy access, greater compliance, and reduced waste
  • A high profile makes the caps easy to handle when wearing gloves
  • Distinctive green color that allows clinicians to verify that a port is protected at a glance
Meeting the high efficacy standards of all Curos™ devices, these caps utilize a spray of 70% isopropyl alcohol (IPA) to disrupt surface microbes, enabling the alcohol to disinfect all critical surfaces of a needleless I.V. valve. Disinfection occurs in only 1 minute and keeps ports clean for 7 days.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty