Would you like to log in again?
Corning® Gosselin™ Round Petri Dish with 6 Vents, Corning
Supplier: Corning
Corning®
BP53-03
75801-452CS 531.56 USD InStock
75801-452
Corning® Gosselin™ Round Petri Dish with 6 Vents, Corning
Culture Dishes Petri Dishes
The Corning® Gosselin™ Round Petri Dish is manufactured from crystal polystyrene, and designed for use in microbiology testing applications
  • Crystal polystyrene construction
  • Double outerbag packaging
  • Easy opening sleeves
  • Sterilized to SAL 10¯6
  • Optimal flatness
The dish is designed with optimal flatness and features six vents. Manufactured in a class 100 environment.
Certifications: Food contact : Raw material is compliant with regulation EC N°1935/2004.
Sterility Assurance Level based on ISO 11137.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty