Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Microvette® 100 K3EDTA Capillary Blood Collection Tube, Sarstedt

Supplier: Sarstedt
Microvette®
20.1278.100
102971-718PK 78.58 USD InStock
102971-718
Microvette® 100 K3EDTA Capillary Blood Collection Tube, Sarstedt
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Microvette® 100 K3EDTA tubes are ideal for capillary blood collection.

  • Equipped with printed label
  • Recommended centrifugation: 2000 x g
  • Shape of inner tube cylindrical
  • Diameter 10,8 mm
  • Length including cap 47,6 mm
  • Length excluding cap 46,6 mm

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty