Would you like to log in again?
XTREME Series Microtubes, Phenix Research
Supplier: Phenix
MAX-815G MAX-705R 1148G43 MAX-815O MAX-705O 1158R24 MAX-820O MAX-805Y MAX-705G MAX-815V MAX-805O MAX-820R
101100-372PK 35.26 USD InStock
101100-372 101100-152 101100-362 101100-176 101100-112 101100-366 101100-356 101100-368 101100-116 101100-358 101100-150 101100-140
XTREME Series Microtubes, Phenix Research
Tubes Microtubes
Microtubes with flat cap and writing surface.
  • Leakproof
  • Writing patch and graduation marks on side
  • Withstands Centrifugation to 20000 ×g
  • Certified RNase/DNase-free and DNA-free
Assorted colors includes blue, green, orange, red, violet, yellow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty