Would you like to log in again?
Mueller Hinton Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
H11
10033-082PK 83.04 USD InStock
10033-082
Mueller Hinton Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For antimicrobial susceptibility testing (AST) of non-fastidious bacteria by utilizing disk diffusion sensitivity method (Kirby-Bauer).
  • Compatible with common BD disk dispensers
  • Broth used for the preparation of suspensions for disk diffusion sensitivity testing
  • Breathable bag packaging prevents build-up of condensation and excess moisture
Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
Packaged in sleeves of 10 plates in a "breathable" bag that prevents build-up of condensation and excess moisture.
Mueller Hinton Agar, Large, for Kirby-Bauer disk susceptibility testing, large 15x150mm plate, order by the package of 10 plates, by Hardy Diagnostics. For antimicrobial susceptibility testing (AST) of non-fastidious bacteria by utilizing disk diffusion sensitivity method (Kirby-Bauer). See HardyDisks AST for a complete selection of Hardy's antibiotic disks for Kirby-Bauer testing. Compatible with common BD disk dispensers. Also see our McFarland standards, dilution broths, and calipers for AST. Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding. Packaged in sleeves of 10 plates in a "breathable" bag that prevents build-up of condensation and excess moisture. The Hardy Diagnostics manufacturing facility and quality management system comply with cGMP's and have achieved ISO 13485 certification as a medical device manufacturer. Mueller Hinton Broth used for the preparation of suspensions for disk diffusion sensitivity testing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty