Would you like to log in again?
Ferguson Angiotribe, OR Grade, Sklar
Supplier: Sklar
17-1260 17-1360
10194-746EA 334.79 USD InStock
10194-746 10194-738
Ferguson Angiotribe, OR Grade, Sklar
Forceps
Ferguson angiotribe forceps are heavy duty scissor shaped hemostats with a screw mechanism, used to control bleeding from a blood vessel.
They are available straight or curved, and in two sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty