Would you like to log in again?
Mycobiotic Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
X30
10324-416PK 166.14 USD InStock
10324-416
Mycobiotic Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of pathogenic fungi.
  • Contains chloramphenicol and cyclohexamide
  • Isolation of fungi from hair, skin, and nail specimens
For the selective isolation of pathogenic fungi especially from hair, skin, and nail specimens. Contains chloramphenicol and cyclohexamide.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty