Would you like to log in again?
MacConkey Agar with Ciprofloxacin, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
G258 G258
10324-446PK 51.41 USD InStock
CA10001-430 10324-446
MacConkey Agar with Ciprofloxacin, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Screening medium for the selective isolation of ciprofloxacin-resistant E. coli and other gram negative rods from clinical samples.
  • Rectal screening medium before they perform a prostate gland biopsy
  • Determine patients at risk of a “difficult to treat” infection or septicemia
  • Selective isolation of ciprofloxacin-resistant E. coli
For use as a screening medium for the selective isolation of ciprofloxacin-resistant E
Urologists use this as a rectal screening medium before they perform a prostate gland biopsy. The biopsy procedure is known for introducing resistant organisms into the bloodstream from the rectum. Our media will allow urologists to screen the men first to see if they will be at risk of a “difficult to treat” infection or septicemia.
coli and other gram negative rods from clinical samples. Especially useful for screening rectal swab specimens prior to prostate gland biopsy. Recommended to be used with a broth enrichment step using BHI with 10µg/ml of ciprofloxacin.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty