Would you like to log in again?
Chocolate Agar Slant, Hardy Diagnostics
Supplier: Hardy Diagnostics
E14BX
89501-516PK 228.65 USD InStock
89501-516
Chocolate Agar Slant, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the non-selective cultivation of fastidious bacteria
  • Highly enriched blend of GC Agar Base
  • Non-selective cultivation of fastidious bacteria
  • Easy to open without a cutting tool
Contains a highly enriched blend of GC Agar Base, 1% bovine powdered hemoglobin, and 1% MaXV (koenzyme) vitamins and cofactor supplements.
Especially useful for the cultivation of Haemophilus and Neisseria in a CO2 enriched atmosphere.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty