Would you like to log in again?
Rapid Anginosus ID Kit, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z14 Z14
10324-562EA 122.01 USD InStock
10324-562 CA10001-518
Rapid Anginosus ID Kit, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the rapid identification of the Streptococcus anginosus group.
  • Results in as little as four hours
  • Easy color change readout
  • Detection of arginine decarboxylase activity
For the rapid identification of the Streptococcus anginosus group (formerly S. milleri) by the detection of arginine decarboxylase activity and production of acetoin. Results in as little as four hours. Easy color change readout, time saving kit format is cost effective. Each kit contains 10 tubes of Rapid Arginine and 10 tubes of Rapid VP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty