Would you like to log in again?
Rapid Urea Medium, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z54 Z54
10324-576PK 72.35 USD InStock
CA10001-524 10324-576
Rapid Urea Medium, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Rapid Urea Medium for rapid detection of urease activity in Helicobacter pylori, Cryptococcus neoformans, or Proteus spp.
  • Positive reactions noted within minutes
  • Rapid detection of urease activity
Rapid Urea Medium for rapid detection of urease activity in Helicobacter pylori, Cryptococcus neoformans, or Proteus spp. Biopsy specimen from endoscopy can be submerged into the agar, positive reactions (pink-red color) may be noted within minutes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty