Would you like to log in again?
Scotchcast™ Wet or Dry Cast Padding, 3M™
Supplier: 3M
Scotchcast™
MMMWDP6 MMMWDP2 MMMWDP4 MMMWDP3
75786-404CS 903.36 USD InStock
75786-404 75786-402 75786-400 75786-398
Scotchcast™ Wet or Dry Cast Padding, 3M™
First Aid Supplies First Aid Kit Components
This synthetic cast padding allows patients to get their cast wet, when following the instructions for use.
  • Sheds water and feels comfortably dry to patient in approximately 1 to 3 hours
  • Suitable for use with all synthetic casting materials
Designed for use in the treatment of muscloskeletal injuries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty