Would you like to log in again?
ClickSpeed™ Magnetic Frame, Vileda Professional®
138621 138695
10153-208CS 777.07 USD InStock
10153-208 10153-216
ClickSpeed™ Magnetic Frame, Vileda Professional®
Mops
The ClickSpeed™ Magnetic Frame is a unique and patented mop frame that is very easy to handle and use.
  • Easily assembled
  • No pockets or Velcro to trap dirt and bacteria
  • 'No Hands' mop attaching and removing
  • Easy pre-preperation of mops
  • Microfiber allows for excellent cleaning results
  • Durable mop is launderable up to 300+ washings
  • Lower water consumption and chemicals needed
ClickSpeed Durable/Reusable Mop Refills are lightweight, knitted, high-performance microfiber mops with polyester stripes. ClickSpeed disposable cloth is a microfiber based cloth with excellent cleaning results. Mop refills are durable in-use and can be washed.
Simply drop the mop on the floor, slide in the frame, and then click-close the magnetic mop holder. This results in touch-free handling of dirty mops, which is beneficial to the user's hygiene.
Ordering information: ClickSpeed Disposable Mop Cloths are packaged 50 per pack/6 packs per case. ClickSpeed Reuseable Mop Cloths are packaged 5 per pack/10 packs per case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty