Would you like to log in again?
UltraSpeed Pro Mop Pads, Vileda Professional-FHP
Supplier: Vileda Professional
143222 147475 143202 143210
75849-354CS 639.2 USD InStock
75849-354 10153-184 10169-892 10169-894
UltraSpeed Pro Mop Pads, Vileda Professional-FHP
Mops
The UltraSpeed Pro system was developed to be simple to use, yet flexible enough to clean difficult areas and offer maximum cleaning performance while significantly reducing mopping times. The result is a comprehensive floor cleaning system that sets new standards in usability, versatility, efficiency, and ergonomics. The UltraSpeed microfiber mop pads are machine washable and easy to use.
  • Lite Version of the MicroPlus™, offering high cleaning intensity with microfiber
  • Polyamide stripes easily clean stubborn dirt
  • Dark grey striped cleaning zones scrub stubborn dirt easily
  • Ideal for structured floors
  • For wet and damp mopping, can also be used for waxing
  • Washable over 300 times
  • Unique color coding tabs (Blue, Red, Green, Yellow)
The MicroPlus microfiber mop pad offers excellent cleaning performance and durability.
The UltraSpeed Trio Mop Pad offers three distinct cleaning zones. The White zone is microfiber for high mechanical cleaning action, and minimizes chemical use. The Grey zone is polyester and provides low friction and high durability. The Beige zone is blended polyester and viscose to provide absorbency. This mop pad has a maximum wash temperature of 95°C (203°F).
The MicroLite Mop Pad is the "lite version" of the series, offering high cleaning intensity with microfiber. Polyamide stripes easily clean stubborn dirt. Ideal for wet and moist mopping, it can also be used for waxing. This mop pad has a maximum wash temperature of 68°C (155°F).
The MicroSafe Mop Pad is a specialty pad designed for cleaning gritty, rough, and safety flooring, and for easily removing stubborn dirt on textured or rough floors. It provides high cleaning performance due to it's abrasive construction to scrub and release dirt, as well as a microfiber component that absorbs and removes dirt. This construction also provides high chemical resistance against chlorine, bleach, and other chemicals required in the cleaning process. The maximum wash temperature for this pad is 70°C (155°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty