Would you like to log in again?
SabHI Agar with Blood, Chloramphenicol and Cycloheximide HardyFlask™, Hardy Diagnostics
Supplier: Hardy Diagnostics
X73
75813-970PK 112.24 USD InStock
75813-970
SabHI Agar with Blood, Chloramphenicol and Cycloheximide HardyFlask™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
SabHI Agar with Blood, Chloramphenicol and Cycloheximide is used for the selective isolation of pathogenic fungi.
  • Selective isolation of pathogenic fungi
  • 5% sheep blood, chloramphenicol and cycloheximide
Sabouraud Dextrose and Brain Heart Infusion Agar with 5% sheep blood, chloramphenicol and cycloheximide, for the selective isolation of pathogenic fungi.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty