Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

epi-Testosterone

Supplier: Pfaltz & Bauer
Warning
Synonyms: Epitestosteroneiso-Testosterone17α-hydroxy-androst-4-en-3-oneNSC 26499

E01160-100MG
101186-748EA 1321.82 USD InStock
101186-748
epi-Testosterone
epi-Testosterone


Formula: C₁₉H₂₈O₂
MW: 288.42 g/mol
Melting Pt: 215…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067129
CAS Number: 481-30-1
Order Now
Specification Test Results
Synonyms 17a-Hydroxy-4-androsten-3-one; 4-Androsten-17a-ol-3-one; 17-epi-Testosterone; 17a-Testosterone; 17a-cis-Testosterone; cis-Testosterone; Isotestosterone; delta-Androsten-17a-ol-3-one

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty