Would you like to log in again?
Steri-Strip™ Antimicrobial Skin Closures, 3M™
Supplier: 3M
Steri-Strip™
MMMA1847 MMMA1840 MMMA1848 MMMA1841 MMMA1842
75787-248CS 1015.23 USD InStock
75787-248 75788-080 75788-108 75788-084 75788-100
Steri-Strip™ Antimicrobial Skin Closures, 3M™
First Aid Supplies Wound Dressings
Closures are made of a porous, non-woven backing coated with a pressure-sensitive, hypoallergenic adhesive and reinforced with filaments for added strength.
Iodophor in the adhesive controls skin bacteria for up to 72 h.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty