Would you like to log in again?
GenePORTER® Gold Transfection Reagent, Genlantis
Supplier: Genlantis
GenePORTER®
T204001S T204001 T204030 T204015 T204115
10497-808EA 4204.46 USD InStock
10497-808 10497-806 10497-804 10497-802 10497-800
GenePORTER® Gold Transfection Reagent, Genlantis
Transfection Reagents
GenePORTER® Gold Transfection Reagent is a highly effective and reproducible lipid based gene delivery formulation for the widest variety of cell lines, especially the easy-to-transfect cell lines.
  • Optimized for same-day transfection
  • Effective in a broad range of cell lines
  • Ideal for high-throughput applications or large scale applications
  • Easiest, and most cost effective transfection
GenePORTER® Gold Transfection Reagent is a highly effective and reproducible lipid based gene delivery formulation for the widest variety of cell lines, especially the easy-to-transfect cell lines. It utilizes the Advanced Carrier Enhancement (ACE) technology that maximizes transfection efficiency and transgene expression levels with minimal cytotoxicity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty