Would you like to log in again?
GeneSilencer® siRNA Transfection Reagent, Genlantis
Supplier: Genlantis
GeneSilencer®
T500750 T505750 T500020 T500250 T500250S
10497-928EA 35.53 USD InStock
10497-928 10497-926 10497-924 10497-932 10497-930
GeneSilencer® siRNA Transfection Reagent, Genlantis
Transfection Reagents
GeneSilencer® siRNA Transfection Reagent is a novel cationic lipid formulation specifically designed for efficient delivery of siRNAs (small interfering RNAs) into a wide variety of cell types.
  • High siRNA transfection efficiency and low cytotoxicity
  • Functional gene silencing post siRNA delivery
  • Compatibility with diverse growth conditions (with and without serum)
  • Stable at 4 °C for six months
  • Easy-to-use protocols for both adherent and suspension cells
GeneSilencer® siRNA Transfection Reagent is a novel cationic lipid formulation specifically designed for efficient delivery of siRNAs (small interfering RNAs) into a wide variety of cell types. siRNAs are short, gene-specific double-stranded RNAs that can cause gene silencing in mammalian cells by catalytically cleaving greater than 95% of the target mRNA (1,2,3).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty