Would you like to log in again?
Tour-Guard™ V Protective Eyewear, 3M™
Supplier: 3M
Tour-Guard™
50051141563911 50051141563904
10837-156CS 403.2 USD InStock
10837-156 10837-158
Tour-Guard™ V Protective Eyewear, 3M™
Safety Glasses
Tour-Guard™ V Protective Eyewear is designed for site visitors or general use.
  • Standard and economical
The eyewear offers reliable impact protection. Intergral sideshields and flexible, polycarbonate construction with over-the-glass coverage. All lenses offer 99.9% UV protection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty