Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Welding Fume Respirator, 3M™
Supplier: 3M
7000002097
89066-612CS 1156.62 USD InStock
89066-612
Welding Fume Respirator, 3M™
Respirators Reusable Respirators Half Face Respirators
Designed for applications including welding, brazing, soldering, torch cutting, and metal pouring, as well as grinding or sanding.
Respirator features a Cool Flow™ exhalation valve, cake-resistant filter media, nose foam, and fully adjustable noseclip and straps.
Certifications: AS/NZS1716 approved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty