Would you like to log in again?
NeuroPORTER® Transfection Reagent, Genlantis
Supplier: Genlantis
NeuroPORTER®
T40010D T400750 T400150 T400100S
10497-826EA 1129.77 USD InStock
10497-826 10497-824 10497-822 10497-922
NeuroPORTER® Transfection Reagent, Genlantis
Transfection Reagents
NeuroPORTER® Transfection Reagent is a novel cationic lipid specially formulated for optimal transfection in neuronal cells, including primary neurons, differentiated post-mitotic neurons, neuronal cell lines, and glial cells.
  • High-level transfection efficiencies in neuronal cell lines, glial cells, and certain primary neurons
  • Optimized protocols for primary neurons, neuronal cell lines, and glial cells
  • Minimized cytotoxicity
NeuroPORTER® Transfection Reagent is compatible with serum eliminates the need to change media following transfection
Storage: Store all reagents at 4 °C. The dried NeuroPORTER reagent is stable for 12 months and the hydrated NeuroPORTER reagent is stable for six months. The Hydration Buffer and DNA Diluent are stable for 12 months.
An included DNA Diluent is designed to facilitate DNA/lipid complex (lipoplex) formation and enhance the transformation efficiency in certain neuronal cells such as NT2 (not recommended for primary and differentiated neurons).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty