Would you like to log in again?
NeuroPure™ E18 Primary Rat Striatal Neuronal Cells, Genlantis
Supplier: Genlantis
N500200
10497-538EA 859.8 USD InStock
10497-538
NeuroPure™ E18 Primary Rat Striatal Neuronal Cells, Genlantis
Cells
NeuroPure™ E18 primary rat striatal cells are live neurons provided as micro-surgically dissected regions of day 18 embryonic sprague/dawley rat brain.
  • Freshly isolated healthy neurons
  • Ready to use eith primary culture up and running within 1 hour
  • Pure neuronal cells - 99.9% glial cell free
Rat striatal cells are prepared fresh each week and shipped in a nutrient rich medium that keeps them alive for up to 6 days under refrigeration. Following simple dissociation steps, the NeuroPure cells can be quickly plated on almost any poly-lysine coated substrate using the provided NeuroPure plating medium. NeuroPure cells are ideal for a wide variety of applications including: transfection, pharmacology, electrophysiology, immunocytochemistry and neuronal development studies.
Delivery information: Includes day 18 Embryonic Sprague/Dawley Rat Striatal Cells ~1 x 106 cells, NeuroPure™ Plating Medium 1 vial x 12 mL, NeuroPrep™ Medium 1 vial x 2.5 mL, NeuroPapain™ Enzyme 1 vial x 5 mg.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty