Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Surgi-OR™ Halsey Needle Holders, Physician Grade, Sklar
Supplier: Sklar
95-810
89023-726EA 52.42 USD InStock
89023-726
Surgi-OR™ Halsey Needle Holders, Physician Grade, Sklar
Forceps
Surgi-OR™ Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology
Surgi-OR™ products are reliable instrumentation for the operating room and other surgical centers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty