Would you like to log in again?
Micro Adson Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
98-2057 98-2047 98-2067
94002-778EA 127.88 USD InStock
94002-778 75996-108 75995-848
Micro Adson Tissue Forceps, OR Grade, Sklar
Forceps
Adson Micro Tissue Forceps are for holding and manipulating delicate tissues
The jaws are short and the tips are narrow. Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures
These forceps have a wide, flat thumb grasp area that is commonly serrated
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty