Would you like to log in again?
Porcine Serum, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SP30-0500HI PS05-0500 SP30-0500 SP30-0100 PS05-0100 SP32-0100
103219-630EA 89.95 USD InStock
103219-630 103219-058 103219-632 103219-056 103219-628 103219-634
Porcine Serum, Equitech- Bio, Inc.
Sera (Serum)
Collected from animals aged 6 months or older.
  • Sterility tested
  • Endotoxin and mycoplasma testing available
  • Lot to lot consistency
Raw serum is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Serum from specific breeds are available by request. Sterile-Filtered serum is sterility tested and sterile-filtered to 0.2 micron absolute. Endotoxin and mycoplasma testing available. Heat-inactivation is available if desired.
Certifications: Certified U.S. origin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty