Would you like to log in again?
Plasmodium falciparum, Gametocytes , Thick Smear Slide
Supplier: Avantor
WARD470181-954
470181-954EA 9.9 USD InStock
470181-954
Plasmodium falciparum, Gametocytes , Thick Smear Slide
Slides Prepared Slides
Can Cause Malaria in Humans
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Microscope slide showing the parasitic protozoan Plasmodium falciparum in human blood.
To learn more about this organism, please visit the CDC's article about the stages of Plasmodium falciparum in blood: https://www.cdc.gov/dpdx/resources/pdf/benchAids/malaria/Pfalciparum_benchaidV2.pdf
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty