Would you like to log in again?
Sklarhone™ Jameson Scissors, OR Grade, Sklar
Supplier: Sklar
15-7032
75817-760EA 284.63 USD InStock
75817-760
Sklarhone™ Jameson Scissors, OR Grade, Sklar
Scissors
Sklarhone™ Jameson Scissors feature curved blades with sharp points used for dissecting or cutting fine tissue
They may also be used for dissecting plaque from an artery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty