Would you like to log in again?
Sklarlite XD™ Halsey Needle Holders, OR Grade, Sklar
Supplier: Sklar
23-2230
75817-772EA 433.15 USD InStock
75817-772
Sklarlite XD™ Halsey Needle Holders, OR Grade, Sklar
Forceps
Sklarlite XD™ Halsey Needle Holders are used for holding small needles for suturing, most commonly during plastic surgery and procedures with delicate tissue, such as dermatology
They are available with smooth or delicately serrated jaws. Smooth jaws will cause the least damage to the needle, however serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Sklarlite XD™ instruments are noticeably lighter and more delicate than standard instruments, allowing the surgeon the highest degree of precision with the least amount of fatigue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty