Would you like to log in again?
Sklarlite XD™ Rochester-Oschner Forceps, OR Grade, Sklar
Supplier: Sklar
23-2577
75817-774EA 187.82 USD InStock
75817-774
Sklarlite XD™ Rochester-Oschner Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Rochester-Oschner Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels
The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps. Sklarlite XD™ instruments are noticeably lighter and more delicate than standard instruments, allowing the surgeon the highest degree of precision with the least amount of fatigue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty