Would you like to log in again?
Nitric Oxide Synthase (NOS) Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB211084-100TEST
75832-948EA 804.65 USD InStock
75832-948
Nitric Oxide Synthase (NOS) Activity Assay Kit (Fluorometric), BioVision
Assays Protein Assays
Ultrasensitive, Simple, HTP adaptable fluorometric assay kit for NOS activity measurement.
  • Can detect as low as 0.75 µU of NOS activity
  • Detection of NOS activity
  • Simple, sensitive and high-throughput adaptable
Nitric oxide synthases (EC 1.14.13.39) (NOS) are a family of enzymes that catalyze the production of nitric oxide (NO) from L-arginine. Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. In presence of NADPH, FAD, FMN, (6R)-5,6,7,8-tetrahydrobiopterin, calmodulin and heme, NOS catalyzes a five-electron oxidation of the guanidino nitrogen of L-arginine with molecular oxygen to generate NO and L-citrulline. There are three isoforms of NOS: endothelial (eNOS), neuronal (nNOS), and inducible (iNOS). nNOS accounts for the production of NO in central nervous system, where NO participates in cell communication and information storage. eNOS produces NO in blood vessels and is involved in regulation of vascular function. In contrast to other isoforms, iNOS is expressed de novo under oxidative stress conditions and produces large amounts of NO as a part of body’s defense mechanism. BioVision’s Nitric Oxide Synthase Activity Assay Kit provides an accurate and convenient method to assay NOS activity in a variety of samples. In this assay, nitric oxide generated by NOS undergoes a series of reactions and reacts with the fluorescent probe to generate a stable signal at Ex/Em = 360/450 nm, which is directly proportional to NOS activity. The assay is simple, sensitive and high-throughput adaptable and can detect as low as 0.75 µU of NOS activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty